Learn More
Chemglass Life Sciences Connection, Complete, GL 25
SDP
Catalog No. CG35410
Encompass_Preferred
Connection, Complete, GL 25. Connector allows for easy connection of a thermocouple gauge tube to glass vacuum system without the use of a glass to metal graded seal or Cajon fitting. Gauge connection consists of a GL 25 thread, a stainless steel bushing and an open top GL 25 screw cap. The thermocouple gauge tube is threaded into the #303 stainless steel bushing and inserted into the glass GL 25 thread.
Catalog No. CG35410 Supplier Chemglass Life Sciences Supplier No. CG35410
Connection, Complete, GL 25. Connector allows for easy connection of a thermocouple gauge tube to glass vacuum system without the use of a glass to metal graded seal or Cajon fitting. Gauge connection consists of a GL 25 thread, a stainless steel bushing and an open top GL 25 screw cap. The thermocouple gauge tube is threaded into the #303 stainless steel bushing and inserted into the glass GL 25 thread.