Learn More
McMaster-Carr Thermocouple Connectors
Catalog No. NC1457109
Encompass
The MCM Thermocouple Connectors are used to connect thermocouples to instruments that measure temperature. They feature nickel-plated brass construction and are available in mini-plug, submini-plug, and standard size plug styles. The mini-plug connectors have a 0.093" diameter plug and fit wire sizes 30-20 AWG. The submini-plug connectors have a 0.070" diameter plug and fit wire sizes 36-28 AWG. The standard connectors have a 0.125" diameter plug and fit wire sizes 24-20 AWG. Operating temperature range is -65 to +165°C. These thermocouple connectors provide a reliable connection for accurate temperature measurements in industrial applications.
Catalog No. NC1457109 Supplier McMaster-Carr Supplier No. 3869K55
The MCM Thermocouple Connectors are used to connect thermocouples to instruments that measure temperature. They feature nickel-plated brass construction and are available in mini-plug, submini-plug, and standard size plug styles. The mini-plug connectors have a 0.093" diameter plug and fit wire sizes 30-20 AWG. The submini-plug connectors have a 0.070" diameter plug and fit wire sizes 36-28 AWG. The standard connectors have a 0.125" diameter plug and fit wire sizes 24-20 AWG. Operating temperature range is -65 to +165°C. These thermocouple connectors provide a reliable connection for accurate temperature measurements in industrial applications.