Learn More
Chemglass Life Sciences Thermocouple Only, Hastings Style, DV-6M
SDP
Catalog No. 50872587
Encompass_Preferred
Thermocouple Only, Hastings Style, DV-6M. Replacement thermocouple gauge tubes for AF-0301-01, AF-0301-02 or AF-0303-01.
Catalog No. 50-872-587 Supplier Chemglass Life Sciences Supplier No. AF030202
Thermocouple Only, Hastings Style, DV-6M. Replacement thermocouple gauge tubes for AF-0301-01, AF-0301-02 or AF-0303-01.