Learn More
J Kem Scientific Inc THERMOCOUPLE PROBES
SDP
Catalog No. NC9880508
Encompass
Teflon-coated type T probe 1/8” diameter x 12” long
Catalog No. NC9880508 Supplier J Kem Scientific Inc Supplier No. T1/8T12
Teflon-coated type T probe 1/8” diameter x 12” long