Learn More
Lascar Electronics Inc Thermocouple Temperature Data Logger with USB
SDP
Catalog No. NC1856606
Encompass_Preferred
This standalone data logger measures over 32,000 readings from inter-changeable J, K or T type thermocouples. Supplied with a K Type probe, its measurement range out of the box is from 0 to 200°C (32 to 392°F). Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery, K-type probe and wall mount clip.
Catalog No. NC1856606 Supplier Lascar Electronics Inc Supplier No. ELUSBTC
This standalone data logger measures over 32,000 readings from inter-changeable J, K or T type thermocouples. Supplied with a K Type probe, its measurement range out of the box is from 0 to 200°C (32 to 392°F). Easily set up the logger and view downloaded data by plugging the unit into a PC’s USB port and using the free EasyLog software provided. Data can then be graphed, printed and exported to other applications for detailed analysis. Supplied with ½ AA battery, K-type probe and wall mount clip.