Learn More
Chemglass Life Sciences Plain Hastelloy C-276 Thermocouple, 1/8" Diameter x 4" Long, Type "K", 4ft FEP Insulated Cord with Standard Male Plug (OST)
SDP
Supplier:  Chemglass Life Sciences CG349830
Encompass_Preferred
Plain Hastelloy C-276 Thermocouple, 1/8" Diameter x 4" Long, Type "K", Standard Male Plug (OST). Thermocouples for use with Chemglass process reactors. Available in both ¿T¿ (blue connections) and ¿K¿ (yellow connections). The detachable thermocouples are for direct insertion in the reactors using a CG-1048-A or CG-1042-E thermocouple adapter, while the 4¿ long Hastelloy C-276 style is for use with the CG-1971 baffles and adapters. Detachable type thermocouples have a standard male plug (OST) and require a coiled extension cord for connection to the CG-3498 temperature monitor. The 4¿ long Hastelloy C-276 thermocouples are supplied with a 4ft FEP insulated cord having a standard male plug (OST).PLEASE NOTE: THE 1/2¿ DIAMETER PROBES HAVE A TRANSITION TO 6¿ OF PTFE LEADS AND ARE CLEAR FEP PTFE COATED.Maximum Probe Temperature Based on Coating- FEP is 200¿C and the Black PFA is 250¿C
Catalog No. CG349830