Learn More
McMaster-Carr Thermocouple Probe for Liquids and Gases, Bendable, with Cable, Type K, Flat Pin Connection, 18" Length Probe.
Catalog No. NC1147335
Encompass
Thermocouple Probe for Liquids and Gases, Bendable, with Cable, Type K, Flat Pin Connection, 18" Length Probe.
Catalog No. NC1147335 Supplier McMaster-Carr Supplier No. 39095K55
Thermocouple Probe for Liquids and Gases, Bendable, with Cable, Type K, Flat Pin Connection, 18" Length Probe.