Learn More
Huber USA Thermofluid DW-Therm M90.200.02
Catalog No. NC0959410
Encompass_Preferred
Unistat-thermofluid for closed systems‐ Temperature min.: -90°C ‐ Temperature max.: 200°C - Viscosity at +25°C: 2mm²/s- Can: 10ltr
Catalog No. NC0959410 Supplier Huber USA Supplier No. 6479
Unistat-thermofluid for closed systems‐ Temperature min.: -90°C ‐ Temperature max.: 200°C - Viscosity at +25°C: 2mm²/s- Can: 10ltr