Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Thermogreen™ LB-1 Septa, Cylindrical

Catalog No. 111020852 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric) Outer:
6 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer
11-102-0852 6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0852 Supplier MilliporeSigma™ Supelco™ Supplier No. 20668
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in glass gas sampling bulbs, and purge & trap glassware

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Outer 6 mm
No. per Pack 100/Pk.
Style Cylindrical with Half Hole
Product Line Thermogreen
For Use With (Equipment) Glass Gas Sampling Bulbs, and Purge ∓ Trap Glassware
Height (Metric) 9 mm
Product Type Septa, LB-1

Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.