Learn More
Perkin Elmer US LLC PerkinElmer Injection Port Septa, Pkg. 50
Catalog No. 509051900
Encompass_Preferred
PerkinElmer Injector Septa, low bleed over a wide range of inlet temperatures -100 to 350 °C. Easier needle penetration and high puncture tolerance make this septum ideal for autosamplers. This septum is already conditioned and ready to use.
Catalog No. 50-905-1900 Supplier Perkin Elmer US LLC Supplier No. N6621028
PerkinElmer Injector Septa, low bleed over a wide range of inlet temperatures -100 to 350 °C. Easier needle penetration and high puncture tolerance make this septum ideal for autosamplers. This septum is already conditioned and ready to use.