Learn More
MilliporeSigma™ Supelco™ Thermogreen™ LB-2 Septa, Pre-drilled
Catalog No. 111020828 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric) Outer:
11 mm
No. per Pack:
50/Pk.
Catalog No. Diameter (Metric) Outer No. per Pack
11-102-0828 11 mm 50/Pk.
1 options
Catalog No. 11-102-0828 Supplier MilliporeSigma™ Supelco™ Supplier No. 23168
Add to Cart
Add to Cart
For use with SPME applications to reduce septum coring
For use with:
  • Minimizing extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS
  • Avoiding carrier gas leaks, extraneous peaks, and phthalate contamination in GC instruments
  • Use in conjunction with a thick needle protector to avoid damage to GC instruments irreversibly caused by microsyringes
  • Influent sampling ports in gas chromatographs
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 7/16 in.
Diameter (Metric) Outer 11 mm
For Use With (Application) SPME
No. per Pack 50/Pk.
Style Pre-drilled
Product Line Thermogreen
Product Type Septa, LB-2
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC