Learn More
Cole-Parmer Extech 445815 Big Digit Thermohygrometer with Dew Point Point, Alarm and Remote Probe
Catalog No. 50993038 Shop All Cole-Parmer Products
Encompass
Designed to monitor relative humidity, and temperature. Features include a super large display, C/F switchable, Min/Max with reset function, low battery indication and a built-in stand for sitting on a desk and hardware for mounting on a wall.
Catalog No. 50-993-038 Supplier Cole-Parmer Supplier No. 3780326
Designed to monitor relative humidity, and temperature. Features include a super large display, C/F switchable, Min/Max with reset function, low battery indication and a built-in stand for sitting on a desk and hardware for mounting on a wall.