Learn More
Cole-Parmer Oakton 7542-00 Thermohygrometer with Digital Thermometer
Supplier:  Cole-Parmer 3570000
Encompass_Preferred
Easily mount this thermohygrometer in your office, laboratory, greenhouse, or food storage environments. This thermohygrometer remains accurate and stable without maintenance or adjustments. Case is constructed of corrosion-resistant ABS plastic. What's included: battery, mounting bracket, and screws.
Catalog No. 50-196-1230