Learn More
New England Biolabs, Inc. Thermolabile Exonuclease I – 3000 units
SDP
Catalog No. NC1698179
Encompass
Thermolabile Exonuclease I catalyzes the removal of nucleotides from single-stranded DNA (ssDNA) in the 3' to 5' direction.
Catalog No. NC1698179 Supplier New England Biolabs, Inc. Supplier No. M0568S
Thermolabile Exonuclease I catalyzes the removal of nucleotides from single-stranded DNA (ssDNA) in the 3' to 5' direction.