Learn More
New England Biolabs, Inc. Thermolabile Exonuclease I – 15000 units
SDP
Catalog No. NC1625476
Encompass
Thermolabile Exonuclease I catalyzes the removal of nucleotides from single-stranded DNA (ssDNA) in the 3' to 5' direction.
Catalog No. NC1625476 Supplier New England Biolabs, Inc. Supplier No. M0568L
Thermolabile Exonuclease I catalyzes the removal of nucleotides from single-stranded DNA (ssDNA) in the 3' to 5' direction.