Learn More
New England Biolabs, Inc. Thermolabile Proteinase K – 30 units
SDP
Catalog No. NC1645675
Encompass
Thermolabile Proteinase K (TLPK) is an engineered, subtilisin-related serine protease that will hydrolyze a broad variety of peptide bonds. It preferentially cleaves the peptide bond at the carboxyl side of aliphatic or aromatic amino acid residues. TLPK can be completely inactivated by incubation at 55°C for 10 minutes, which allows for subsequent enzymatic steps in the same reaction vessel.
Catalog No. NC1645675 Supplier New England Biolabs, Inc. Supplier No. P8111S
Thermolabile Proteinase K (TLPK) is an engineered, subtilisin-related serine protease that will hydrolyze a broad variety of peptide bonds. It preferentially cleaves the peptide bond at the carboxyl side of aliphatic or aromatic amino acid residues. TLPK can be completely inactivated by incubation at 55°C for 10 minutes, which allows for subsequent enzymatic steps in the same reaction vessel.