Learn More
Qiagen Beverly Inc THERMOLABILE UDG
Catalog No. 50305942
Encompass
Glycosylase; Thermolabile Uracil DNA Glycosylase; Conc. 1,000 U/ul; 500U; incl. 10X buffer
Catalog No. 50-305-942 Supplier Qiagen Beverly Inc Supplier No. G5020L
Glycosylase; Thermolabile Uracil DNA Glycosylase; Conc. 1,000 U/ul; 500U; incl. 10X buffer