Learn More
New England Biolabs, Inc. Thermolabile USER II Enzyme – 50 units
SDP
Catalog No. 501526411
Encompass
Thermolabile USER (Uracil-Specific Excision Reagent) II Enzyme generates a single nucleotide gap at the location of a uracil residue. It can be 100% inactivated at temperatures >65C.
Catalog No. 50-152-6411 Supplier New England Biolabs, Inc. Supplier No. M5508S
Thermolabile USER (Uracil-Specific Excision Reagent) II Enzyme generates a single nucleotide gap at the location of a uracil residue. It can be 100% inactivated at temperatures >65C.