Learn More
New England Biolabs, Inc. Thermolabile USER II Enzyme – 250 units
SDP
Catalog No. 501526410
Encompass
Thermolabile USER (Uracil-Specific Excision Reagent) II Enzyme generates a single nucleotide gap at the location of a uracil residue. It can be 100% inactivated at temperatures >65C.
Catalog No. 50-152-6410 Supplier New England Biolabs, Inc. Supplier No. M5508L
Thermolabile USER (Uracil-Specific Excision Reagent) II Enzyme generates a single nucleotide gap at the location of a uracil residue. It can be 100% inactivated at temperatures >65C.