Learn More
Icl Cal Lab Inc NIST Traceable, ISO/IEC 17025 accredited thermometers, -5 to 130 °C, -5 to 130 °C 
SDP
Catalog No. NC0165208
Encompass
Specifically designed to comply with CLIA and EPA specifications. Supplied with NIST Traceable, ISO/IEC 17025 accredited certificate of calibration. Precision mercury in glass or Non Mercury thermometers.Graduated in 0.5 ° Divisions. Please provide calibration interval on each order, 1 or 2 years
Catalog No. NC0165208 Supplier Icl Cal Lab Inc Supplier No. 210008RL
Specifically designed to comply with CLIA and EPA specifications. Supplied with NIST Traceable, ISO/IEC 17025 accredited certificate of calibration. Precision mercury in glass or Non Mercury thermometers.Graduated in 0.5 ° Divisions. Please provide calibration interval on each order, 1 or 2 years