Learn More
Hach Company Thermometer, 0-200° C
Supplier:  Hach Company 4565500
Encompass
Bi-metallic. Dial gauge 0 to 200°C ±1%. Two degree divisions. Probe diameter is .15 inches. Plastic carry case with clip.
Catalog No. 50-199-7045