Learn More
Thermco Products Inc Precision lab Hg thermometer, -10 deg./200 deg.C, 1.0 deg.C div, TOTAL imm, 305mm
Catalog No. NC0605433
Encompass_Preferred
High precision mercury SAMA thermometer that meet or exceed the specifications of the American National Standards Institute. Specifications approve by N.I.S.T. and ASTM. Individually serialized and permanetly marked. Made with precision yellow backed tubing.
Catalog No. NC0605433 Supplier Thermco Products Inc Supplier No. ACC615
High precision mercury SAMA thermometer that meet or exceed the specifications of the American National Standards Institute. Specifications approve by N.I.S.T. and ASTM. Individually serialized and permanetly marked. Made with precision yellow backed tubing.