Learn More
Andwin Scientific THERMOMETER -20/110C
SDP
Catalog No. NC0910946
Encompass_Preferred
The General Purpose Blue Spirit Thermometer can measure within the temperature range of 20 to 110C. It features accurate results of + or - 1.5C. This product has an overall length of 300 mm and an immersion of 76 mm. Each thermometer is individual
Catalog No. NC0910946 Supplier Andwin Scientific Supplier No. 89095598
The General Purpose Blue Spirit Thermometer can measure within the temperature range of 20 to 110C. It features accurate results of + or - 1.5C. This product has an overall length of 300 mm and an immersion of 76 mm. Each thermometer is individual