Learn More
Hach Company Thermometer, Enviro-Safe, Non-Mercury, -20 to 110°C
Supplier:  Hach Company 2635702
Encompass
Enviro-Safe non-Mercury thermometers feature an exclusive biodegradable liquid and dye. Easy to read, aesthetically distinctive due to dark green liquid and permanently fused markings. Each thermometer, individually calibrated against NIST traceable equipment, identified by unique serial number for ISO9000 or other quality systems.
Catalog No. 50-199-6403