Learn More
Thermco Products Inc Precision lab Non-Hg thermometer, Spirit, w/safety coat, -35 deg./50 deg.F, 1 deg.F div, 76mm imm, 305mm
Catalog No. NC0539913
Encompass_Preferred
High precision Spirit Filled thermometer that is accurate to within N.I.S.T.tolerances. Individually serialized and permanetly marked. Made with precision whie backed tubing. Thermometer is coated with PTFE for prtection of breakage. The coating will contain the glass and liquid if breakage occurs. Eligible for N.I.S.T. certification
Catalog No. NC0539913 Supplier Thermco Products Inc Supplier No. ACC6113SSC
High precision Spirit Filled thermometer that is accurate to within N.I.S.T.tolerances. Individually serialized and permanetly marked. Made with precision whie backed tubing. Thermometer is coated with PTFE for prtection of breakage. The coating will contain the glass and liquid if breakage occurs. Eligible for N.I.S.T. certification