Learn More
Chemglass Life Sciences Thermometer, Non-Mercury, Total Immersion, -20°C to 110°C, 305mm OAL
SDP
Catalog No. CG350101
Encompass_Preferred
Thermometer, Non-Mercury, Total Immersion, -20¿C to 110¿C, 305mm OAL. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. All ranges have 1¿ subdivisions. Thermometer has a white back and a 6 to 7mm outer diameter.
Catalog No. CG350101 Supplier Chemglass Life Sciences Supplier No. CG350101
Thermometer, Non-Mercury, Total Immersion, -20¿C to 110¿C, 305mm OAL. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. All ranges have 1¿ subdivisions. Thermometer has a white back and a 6 to 7mm outer diameter.