Learn More
Hach Company Thermometer, Non-mercury, 30 to 50° C
Supplier:  Hach Company 2999200
Encompass
Non-mercury thermometer, 30 - 50 degrees C Length: 250 mm Immersion: 76 mm Safety coated. Blue fill solution on white background provides excellent visibility.
Catalog No. 50-199-7008