Learn More
Thermco Products Inc THERMOMETER
Catalog No. NC1254922
Encompass_Preferred
NIST traceable - Factory Certified Non-Mercury Glass Thermometers - Spirit Filled, -1/51C x 0.1C Partial immersion, Cert @ 0, 10, 20, 30, 37, 40, 50C
Catalog No. NC1254922 Supplier Thermco Products Inc Supplier No. ACC10033SFC
NIST traceable - Factory Certified Non-Mercury Glass Thermometers - Spirit Filled, -1/51C x 0.1C Partial immersion, Cert @ 0, 10, 20, 30, 37, 40, 50C