Learn More
Interworld Highway LLC Tequipment Net Net Fluke 51-2 Thermometer Series II
SDP
Catalog No. NC1377480
Encompass_Preferred
Digital Thermometer, 210 To 1200 Deg C JType, 200 To 1372 Deg C KType, 250 To 400 Deg C TType, 150. Relative time clock on MIN, MAX, and AVG provides a time reference for major events. Electronic Offset function allows compensation of thermocouple errors to maximize overall accuracy. Measures J, K, T, and E-types of thermocouples. Readout in °C, °F, or Kelvin (K). Splash and dust resistant case protected by impact absorbing holster. User-friendly front panel is easy to set up and operate. Sleep mode increases battery life; typical 1000-hour battery life. Battery door allows easy battery replacement without breaking the calibration seal. Optional ToolPak accessory allows the thermometer to hang from any metal object (with the rare earth magnet) or secure around a pipe (with hook-and-loop straps) for hands-free operation
Catalog No. NC1377480 Supplier Interworld Highway LLC Tequipment Net Supplier No. FLUKE 512 60HZ
You must be signed in and have a business account to purchase online.
Digital Thermometer, 210 To 1200 Deg C JType, 200 To 1372 Deg C KType, 250 To 400 Deg C TType, 150. Relative time clock on MIN, MAX, and AVG provides a time reference for major events. Electronic Offset function allows compensation of thermocouple errors to maximize overall accuracy. Measures J, K, T, and E-types of thermocouples. Readout in °C, °F, or Kelvin (K). Splash and dust resistant case protected by impact absorbing holster. User-friendly front panel is easy to set up and operate. Sleep mode increases battery life; typical 1000-hour battery life. Battery door allows easy battery replacement without breaking the calibration seal. Optional ToolPak accessory allows the thermometer to hang from any metal object (with the rare earth magnet) or secure around a pipe (with hook-and-loop straps) for hands-free operation