Learn More
Kessler Thermometer Corp THERMOMETER 38/82C WITH CERT
SDP
Supplier:  Kessler Thermometer Corp 1051SBC
Encompass
Thermometer 38/82C made to the same specifications as ASTM 14C except with safety blue liquid.Certification included 14C38 to 82°C0.1375mm79mmMinimum of 6 each per order
Catalog No. NC3253108