Learn More
Thermco Products Inc Precision lab Non-Hg thermometer, Spirit, Fractional degree, -1 deg./51 deg.C, 0.1 deg.C div, 76mm imm, 460mm
Catalog No. NC0573828
Encompass_Preferred
High precision Spirit Filled thermometer that is accurate to within N.I.S.T.tolerances. Individually serialized and permanetly marked. Made with precision whie backed tubing. Eligible for N.I.S.T. certification
Catalog No. NC0573828 Supplier Thermco Products Inc Supplier No. ACC10033S
High precision Spirit Filled thermometer that is accurate to within N.I.S.T.tolerances. Individually serialized and permanetly marked. Made with precision whie backed tubing. Eligible for N.I.S.T. certification