Learn More
Chemglass Life Sciences Thermometer, Non-Mercury, Partial Immersion, -20°C to 150°C, 305mm OAL
SDP
Catalog No. CG350311
Encompass_Preferred
Thermometer, Non-Mercury, Partial Immersion, -20¿C to 150¿C, 305mm OAL. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. All ranges have 1¿ subdivisions. Thermometer has a white back and a 6 to 7mm outer diameter, and is calibrated for 76mm (3") immersion.
Catalog No. CG350311 Supplier Chemglass Life Sciences Supplier No. CG350311
Thermometer, Non-Mercury, Partial Immersion, -20¿C to 150¿C, 305mm OAL. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. All ranges have 1¿ subdivisions. Thermometer has a white back and a 6 to 7mm outer diameter, and is calibrated for 76mm (3") immersion.