Learn More
Braintree Scientific Hand Held Temperature Measurement w/Time Measurement
SDP
Catalog No. NC1534388
Encompass
Bioseb Hand Held Temperature Measurement. With time measurement. An ideal tool for all your temperature measurements on rodents (rats or mice) which features a large backlit digital display and time measurement.Features-- 1 T1 channel Resolution: 0,1C. Functions: records max, records min, hold 2.5 measurements/sec refreshing screen Time measurement display from start with dedicated key.Supplied with-- Black plastic box Batteries Rectal probe not included in the price - accepts all type "T" thermocouple probes **Sold for research purposes only and not for human use**
Catalog No. NC1534388 Supplier Braintree Scientific Supplier No. TK8851
Bioseb Hand Held Temperature Measurement. With time measurement. An ideal tool for all your temperature measurements on rodents (rats or mice) which features a large backlit digital display and time measurement.Features-- 1 T1 channel Resolution: 0,1C. Functions: records max, records min, hold 2.5 measurements/sec refreshing screen Time measurement display from start with dedicated key.Supplied with-- Black plastic box Batteries Rectal probe not included in the price - accepts all type "T" thermocouple probes **Sold for research purposes only and not for human use**