Learn More
OHAUS™ Thermometer Clamp
Catalog No. 01922034 Shop All OHAUS Products
Change view
Click to view available options
Size (Metric) Grip:
6 mm - 12 mm
6 mm - 8 mm
Type:
Thermometer Clamp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Grip Type
01-922-034 6 mm - 8 mm Thermometer Clamp
01-922-035 6 mm - 12 mm Thermometer Clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-922-034 Supplier OHAUS™ Supplier No. 30392238
Add to Cart
Add to Cart
Multipurpose clamps
  • LabJaws Clamps are designed to securely grip and position laboratory apparatus.
  • Extension arm clamps with 360° rotation allow placement of apparatus at various distances from lab-frames and are easily secured with optional holders.
  • Swivel, fixed position, and thermometer clamps easily hold apparatus near the lab-frame or stand, and have a built-in holder for simple attachment to lab-frames or apparatus.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Chromatography, Distillation, Titrations, Electrolysis, Chemistry, Research & Development, Quality Control, Ring Stands, Water Baths, Cleanroom Applications, Acid Fume Hood Applications
Model CLS-THMSWZ
Type Thermometer Clamp
Material Nickel-plated Zinc
Size (Metric) Grip 6 mm - 8 mm