Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Thermometer Clamp Extension
GREENER_CHOICESDP

Catalog No. 31501958 Shop All Kemtech Products
Change view
Click to view available options
Length (Metric):
200 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
31-501-958 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-501-958 Supplier Synthware™ Supplier No. W140200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Extension rod 10 mm in diameter. Total length: 200 mm.

Specifications

Type Thermometer Extension Clamp
Material Metal
For Use With (Application) Organic Synthesis
Diameter (Metric) 10 mm
Length (Metric) 200 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.