Learn More
Hach Company Digital Thermometer
Supplier:  Hach Company 2630600
Encompass
Light weight and inexpensive, this K & J type thermometer provides extremely accurate and reliable temperature measurement. Features relative minimum and maximum temperature, hold, auto power off, and touch tone buttons. This is the meter only. For use with probe 2630601 (sold seprately).
Catalog No. 50-199-6395