Learn More
Cole-Parmer Fluke 54-2 B 60HZ Dual-Input Datalogging Thermocouple Thermometer
Catalog No. NC9334797 Shop All Cole-Parmer Products
Encompass
Dual backlit display provides readout of channel temperature, differential temperature (dual-input models), and minimum/maximum/average readings with time stamp.
Catalog No. NC9334797 Supplier Cole-Parmer Supplier No. 9150010
Customers require a valid business account associated to their profile to access pricing and ordering options.
Dual backlit display provides readout of channel temperature, differential temperature (dual-input models), and minimum/maximum/average readings with time stamp.