Learn More
Med Vet International Medline Talking Ear And Forehead infrared Thermometer, Each
Catalog No. 502831986
Encompass_Preferred
MEDLINE Thermometer Talking Ear Forehead Infrared MDSTH1002 New Unopened Fresh. Speaks in 6 languages English, Spanish, French, German, Italian, or Russian. The Medline Digital Talking Ear & Forehead Thermometer accurately measures body heat using Rapid, Accurate, State of the Art Technology. Sensor rapidly tracks the heat flow generated from blood vessels to the skin surface and converts measurement to body temperature. GREAT FOR BABY, TODDLER, CHILD OR ADULT
Catalog No. 50-283-1986 Supplier Med Vet International Supplier No. MDSTH1002
 Restricted Item
Add to Cart
Add to Cart
MEDLINE Thermometer Talking Ear Forehead Infrared MDSTH1002 New Unopened Fresh. Speaks in 6 languages English, Spanish, French, German, Italian, or Russian. The Medline Digital Talking Ear & Forehead Thermometer accurately measures body heat using Rapid, Accurate, State of the Art Technology. Sensor rapidly tracks the heat flow generated from blood vessels to the skin surface and converts measurement to body temperature. GREAT FOR BABY, TODDLER, CHILD OR ADULT