Learn More
Thermco Products Inc Stainless Thermometer/pH Electrode/Test Tube Holder
Catalog No. NC0785726
Encompass_Preferred
Thermometer and pH probe holder with a pan clip. Clips firmly on the side of a beaker. The clip has rubber clamping pads. The holder is foam rubber with 5 holesof assorted sizes.
Catalog No. NC0785726 Supplier Thermco Products Inc Supplier No. ACCTH1000
Thermometer and pH probe holder with a pan clip. Clips firmly on the side of a beaker. The clip has rubber clamping pads. The holder is foam rubber with 5 holesof assorted sizes.