Learn More
Grainger PolyScience Thermometer Holder, For Circulating Baths, 1 ea
Catalog No. NC1146409
Encompass_Preferred
This thermometer holder is designed to securely clip onto equipment, providing stability to ensure accurate temperature readings during operation.
  • Designed for use with circulating baths and open bath systems
  • Secure clip-on mechanism for easy attachment
  • Compatible with stainless steel and polycarbonate bath systems
  • Suitable for specialty products and cryoprecipitate baths
Catalog No. NC1146409 Supplier Grainger Supplier No. 22FH25
This thermometer holder is designed to securely clip onto equipment, providing stability to ensure accurate temperature readings during operation.
  • Designed for use with circulating baths and open bath systems
  • Secure clip-on mechanism for easy attachment
  • Compatible with stainless steel and polycarbonate bath systems
  • Suitable for specialty products and cryoprecipitate baths