Learn More
Chemglass Life Sciences Thermometer Holder, Polypropylene
SDP
Catalog No. 501311224
Encompass_Preferred
Thermometer Holder, Polypropylene. This molded polypropylene thermometer holder clips onto beakers and other vessels and can hold a thermometer from 5 to 11mm OD. Sold in packages of 3.
Catalog No. 50-131-1224 Supplier Chemglass Life Sciences Supplier No. CG351101
Thermometer Holder, Polypropylene. This molded polypropylene thermometer holder clips onto beakers and other vessels and can hold a thermometer from 5 to 11mm OD. Sold in packages of 3.