Learn More
Med Vet International Non-Contact Skin Surface Thermometer infrared Skin Probe, Th-300, Each
Catalog No. 502829982
Encompass_Preferred
Fast 1 second response time Large easy to read LCD Display shows temperature in Fahrenheit or Celsius Reads skin surface temperature with infrared technology On/Off power button with automatic shut off
Catalog No. 50-282-9982 Supplier Med Vet International Supplier No. 1167501EA
 Restricted Item
Add to Cart
Add to Cart
Fast 1 second response time Large easy to read LCD Display shows temperature in Fahrenheit or Celsius Reads skin surface temperature with infrared technology On/Off power button with automatic shut off