Learn More
Chemglass Life Sciences Thermometer Magnifier
SDP
Catalog No. 501311223
Encompass_Preferred
Thermometer Magnifier. Thermometer magnifier allows for precise readings of any thermometer from 5 to 9mm OD. The hide a line feature eliminated parallax and magnifies up to 2-1/2 times. Magnifier slides up and down the thermometer.
Catalog No. 50-131-1223 Supplier Chemglass Life Sciences Supplier No. CG350910
Thermometer Magnifier. Thermometer magnifier allows for precise readings of any thermometer from 5 to 9mm OD. The hide a line feature eliminated parallax and magnifies up to 2-1/2 times. Magnifier slides up and down the thermometer.