Learn More
Thermco Products Inc Autoclave max register thermometer, refill, w/safety coat, 80 deg./135 deg.C
Catalog No. NC0228775
Encompass_Preferred
*Metal armor not included for NC0228775* Maximum registering thermometer used for the verification of the temperature in autoclaves and sterilizers. The thermometer will indicate the highest temperature in such instruments. After each use the thermometer must be shaken down to reset the thermometer before being used again. The ACC80135SC is safety-coated with PTFE, so no glass or mercury will be emited if broken. Can be certified treceable to N.I.S.T. individually serialized.
Catalog No. NC0228775 Supplier Thermco Products Inc Supplier No. ACC80135SC
*Metal armor not included for NC0228775* Maximum registering thermometer used for the verification of the temperature in autoclaves and sterilizers. The thermometer will indicate the highest temperature in such instruments. After each use the thermometer must be shaken down to reset the thermometer before being used again. The ACC80135SC is safety-coated with PTFE, so no glass or mercury will be emited if broken. Can be certified treceable to N.I.S.T. individually serialized.