Learn More
Med Vet International Medline Thermometer, Hud1647, 50/Case
Catalog No. 502829802
Encompass_Preferred
Clear housing with "easy read" red liquid column Fahrenheit and Celsius scales from 50 degrees to 120 degrees and 10 degrees to 50 degrees respectively Probe fits most temperature ports and aerosol tee adaptors
Catalog No. 50-282-9802 Supplier Med Vet International Supplier No. 171565CS
Clear housing with "easy read" red liquid column Fahrenheit and Celsius scales from 50 degrees to 120 degrees and 10 degrees to 50 degrees respectively Probe fits most temperature ports and aerosol tee adaptors