Learn More
Med Vet International Medline Thermometer, Hud1647, Each
Catalog No. 502829681
Encompass_Preferred
Clear housing with "easy read" red liquid column Fahrenheit and Celsius scales from 50 degrees to 120 degrees and 10 degrees to 50 degrees respectively Probe fits most temperature ports and aerosol tee adaptors
Catalog No. 50-282-9681 Supplier Med Vet International Supplier No. 171565EA
Clear housing with "easy read" red liquid column Fahrenheit and Celsius scales from 50 degrees to 120 degrees and 10 degrees to 50 degrees respectively Probe fits most temperature ports and aerosol tee adaptors