Learn More
Thermco Products Inc Precision Laboratory Thermometer, Red Spirit Filled, Non-Mercury Fractional Degree, 5, 76mm Partial Immersion, 610mm Length
Catalog No. NC1095218
Encompass_Preferred
Precision Laboratory Thermometer, Red Spirit Filled, Non-Mercury Fractional Degree, 5, 76mm Partial Immersion, 610mm Length. NIST TraceableNon-Mercury Red Spirit Filled Thermometers. Total and Partial Immersion. Individually Serializedrialized.
Catalog No. NC1095218 Supplier Thermco Products Inc Supplier No. ACC10053S
Precision Laboratory Thermometer, Red Spirit Filled, Non-Mercury Fractional Degree, 5, 76mm Partial Immersion, 610mm Length. NIST TraceableNon-Mercury Red Spirit Filled Thermometers. Total and Partial Immersion. Individually Serializedrialized.