Learn More
Chemglass Life Sciences Thermometer, Non-Mercury Low Temperature, Partial Immersion, -100°C to 50°C
SDP
Supplier:  Chemglass Life Sciences CG3503L21
Encompass_Preferred
Thermometer, Non-Mercury Low Temperature, Partial Immersion, -100¿C to 50¿C. Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury. All ranges have 1¿ subdivisions. Thermometer has a white back and a 6 to 7mm outer diameter. Available in total or partial 76mm (3") immersion.
Catalog No. CG3503L21