Learn More
Hach Company Thermometer, Dual Range, Non-Mercury, -10 to 110°C, 0 to 220°F
Catalog No. 501996527 Shop All Hach Company Products
Encompass
Compact and durable, non-mercury thermometers enclosed in a protective nylon case. This thermometer is designed for ruggedness. Suitable for all field and portable uses where an unprotected thermometer may break.
Catalog No. 50-199-6527 Supplier Hach Company Supplier No. 2676400
Compact and durable, non-mercury thermometers enclosed in a protective nylon case. This thermometer is designed for ruggedness. Suitable for all field and portable uses where an unprotected thermometer may break.