Learn More
Cole-Parmer Extech 39272 Adjustable Angle Thermometer
Catalog No. 50993109 Shop All Cole-Parmer Products
Encompass
This thermometer takes fast and accurate measurements anywhere. Tapered probe can be used in solids, semisolids, liquids and gases. Large ½" high display provides great visibility. Fold up 4.5"L (114mm) probe adjusts from 0° to 180° for best measurement and viewing angle. Features data hold and min/max functions. Auto shut-off after one hour of non-use to conserve battery life. What’s included: One AAA battery and wrist strap
Catalog No. 50-993-109 Supplier Cole-Parmer Supplier No. 9038200
This thermometer takes fast and accurate measurements anywhere. Tapered probe can be used in solids, semisolids, liquids and gases. Large ½" high display provides great visibility. Fold up 4.5"L (114mm) probe adjusts from 0° to 180° for best measurement and viewing angle. Features data hold and min/max functions. Auto shut-off after one hour of non-use to conserve battery life. What’s included: One AAA battery and wrist strap